Methyl-6-(dimethylamino)nicotinate 98%
Dodavatel: Apollo Scientific
Synonyms:
Methyl 6-dimethylamino-3-pyridinecarboxylate
OR902009-1G
OR902009-5G
APOSOR902009-1GEA
4770
CZK
APOSOR902009-1G
APOSOR902009-5G
Methyl-6-(dimethylamino)nicotinate 98%
Methyl-6-(dimethylamino)nicotinate
Vzorec:
C₉H₁₂N₂O₂ MW: 180,21 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD10699718 CAS číslo: 26218-81-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...