2-(Ethylamino)-3-nitro-5-picoline 98%
Dodavatel: Apollo Scientific
Synonyms:
2-Ethylamino-5-methyl-3-nitropyridine
OR902004-100G
OR902004-25G
OR902004-5G
APOSOR902004-100GEA
18420
CZK
APOSOR902004-100G
APOSOR902004-25G
APOSOR902004-5G
2-(Ethylamino)-3-nitro-5-picoline 98%
2-(Ethylamino)-3-nitro-5-picoline
Vzorec:
C₈H₁₁N₃O₂ Storage Temperature: Ambient |
MDL Number:
MFCD09862688 CAS číslo: 106690-40-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...