Tisk…

Ethyl-6-chloropicolinate 97%

Dodavatel: Apollo Scientific

Synonyms: Ethyl 6-chloropicolinate, Ethyl 6-chloropyridine-2-carboxylate

OR901965-25G
APOSOR901965-25GEA 2360 CZK
APOSOR901965-25G
Ethyl-6-chloropicolinate 97%
Ethyl-6-chloropicolinate
Vzorec: C₈H₈ClNO₂
MW: 185,61 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09878438
CAS číslo: 21190-89-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR