Tisk…

2-(5-Bromopyridin-2-yl)acetonitrile 97%

Dodavatel: Apollo Scientific
Varování

OR901941-1G OR901941-5G
APOSOR901941-1GEA 1370 CZK
APOSOR901941-1G APOSOR901941-5G
2-(5-Bromopyridin-2-yl)acetonitrile 97%
2-(5-Bromopyridin-2-yl)acetonitrile
Vzorec: C₇H₅BrN₂
Storage Temperature: Ambient
MDL Number: MFCD07644625
CAS číslo: 312325-72-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR