Tisk…

5-Chloro-2-methoxypyridin-3-ol 98%

Dodavatel: Apollo Scientific

OR901850-1G OR901850-250MG OR901850-5G
APOSOR901850-1GEA 9330 CZK
APOSOR901850-1G APOSOR901850-250MG APOSOR901850-5G
5-Chloro-2-methoxypyridin-3-ol 98%
5-Chloro-2-methoxypyridin-3-ol
Vzorec: C₆H₆ClNO₂
Storage Temperature: Ambient
MDL Number: MFCD18374102
CAS číslo: 1261365-86-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR