Tisk…

2-(Diethylamino)nicotinonitrile 95%

Dodavatel: Apollo Scientific

Synonyms: 3-Cyano-2-diethylaminopyridine

OR901731-1G OR901731-5G
APOSOR901731-1GEA 4770 CZK
APOSOR901731-1G APOSOR901731-5G
2-(Diethylamino)nicotinonitrile 95%
2-(Diethylamino)nicotinonitrile
Vzorec: C₁₀H₁₃N₃
MW: 175,24 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09896569
CAS číslo: 17782-02-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR