Tisk…

6-Methyl-2-quinolinol 98%

Dodavatel: Apollo Scientific

Synonyms: 6-Methylquinolin-2(1H)-one

OR901618-1G OR901618-5G
APOSOR901618-1GEA 2030 CZK
APOSOR901618-1G APOSOR901618-5G
6-Methyl-2-quinolinol 98%
6-Methyl-2-quinolinol
Vzorec: C₁₀H₉NO
MW: 159.1846 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08236767
CAS číslo: 4053-34-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR