Tisk…

5-Bromopyridine-3-sulphonyl chloride 97%

Dodavatel: Apollo Scientific
Nebezpečí

OR9015-1G OR9015-250MG
APOSOR9015-1GEA 7970 CZK
APOSOR9015-1G APOSOR9015-250MG
5-Bromopyridine-3-sulphonyl chloride 97%
5-Bromopyridine-3-sulphonyl chloride
Vzorec: C₅H₃BrClNO₂S
Bod tání: 60…63 °C
Storage Temperature: Ambient
MDL Number: MFCD08235263
CAS číslo: 65001-21-0
UN: 2923
ADR: 8+6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR