(3-Tolyloxy)acetic acid 98%
Dodavatel: Apollo Scientific
Synonyms:
2-(m-Tolyloxy)acetic acid, (3-Methylphenoxy)acetic acid
OR900712-25G
APOSOR900712-25GEA
4300
CZK
APOSOR900712-25G
(3-Tolyloxy)acetic acid 98%
(3-Tolyloxy)acetic acid
Vzorec:
(C₆H₄(CH₃))OCH₂COOH MW: 166,18 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00014358 CAS číslo: 1643-15-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...