(2-Tolyloxy)acetic acid 98%
Dodavatel: Apollo Scientific
Synonyms:
2-(o-Tolyloxy)acetic acid, (2-Methylphenoxy)acetic acid
OR900710-100G
OR900710-25G
APOSOR900710-100GEA
13360
CZK
APOSOR900710-100G
APOSOR900710-25G
(2-Tolyloxy)acetic acid 98%
(2-Tolyloxy)acetic acid
Vzorec:
(C₆H₄CH₃)OCH₂COOH MW: 166,18 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00014354 CAS číslo: 1878-49-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...