Tisk…

(4-Tolyloxy)acetic acid 98%

Dodavatel: Apollo Scientific

OR900698-100G OR900698-25G
APOSOR900698-100GEA 9820 CZK
APOSOR900698-100G APOSOR900698-25G
(4-Tolyloxy)acetic acid 98%
(4-Tolyloxy)acetic acid
Vzorec: (C₆H₄(CH₃))OCH₂COOH
MW: 166,18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00014365
CAS číslo: 940-64-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR