Tisk…

3-Chloro-4-methoxyphenol 95%

Dodavatel: Apollo Scientific

OR900665-10G OR900665-1G OR900665-5G
APOSOR900665-10GEA 28540 CZK
APOSOR900665-10G APOSOR900665-1G APOSOR900665-5G
3-Chloro-4-methoxyphenol 95%
3-Chloro-4-methoxyphenol
Vzorec: C₇H₇ClO₂
Storage Temperature: Ambient
CAS číslo: 18093-12-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR