1-(Dimethylamino)cyclohexanecarbonitrile 96%
Dodavatel: Apollo Scientific
OR900600-1G
OR900600-25G
OR900600-5G
APOSOR900600-1GEA
2900
CZK
APOSOR900600-1G
APOSOR900600-25G
APOSOR900600-5G
1-(Dimethylamino)cyclohexanecarbonitrile 96%
1-(Dimethylamino)cyclohexanecarbonitrile
Vzorec:
C₉H₁₆N₂ MW: 152,24 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00086186 CAS číslo: 16499-30-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...