Tisk…

Naphthol yellow S

Dodavatel: Apollo Scientific
Varování

Synonyms: Flavianic acid disodium salt, Disodium 2,4-Dinitro-1-naphthol-7-sulphonate, 8-Hydroxy-5,7-dinitro-2-naphthalenesulphonic acid disodium salt, 2,4-Dinitro-1-naphthol-7-sulphonic acid disodium salt, Disodium 8-Hydroxy-5,7-dinitro-2-naphthalenesulphonate

OR9003-25G OR9003-100G
APOSOR9003-25GEA 2690 CZK
APOSOR9003-25G APOSOR9003-100G
Naphthol yellow S
Naphthol yellow S
Vzorec: C₁₀H₆N₂O₈S
MW: 358,19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003958
CAS číslo: 846-70-8
EINECS: 212-690-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR