4-(Tetrahydropyran-4-yl)phenylamine 98%
Dodavatel: Apollo Scientific
Synonyms:
4-(Oxan-4-yl)aniline
OR900268-1G
OR900268-250MG
APOSOR900268-1GEA
6910
CZK
APOSOR900268-1G
APOSOR900268-250MG
4-(Tetrahydropyran-4-yl)phenylamine 98%
4-(Tetrahydropyran-4-yl)phenylamine
Vzorec:
C₁₁H₁₅NO Storage Temperature: Ambient |
MDL Number:
MFCD11506022 CAS číslo: 62071-40-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...