Tisk…

6-Methoxy-2-naphthaldehyde 98%

Dodavatel: Apollo Scientific

OR900156-100G OR900156-25G
APOSOR900156-100GEA 4960 CZK
APOSOR900156-100G APOSOR900156-25G
6-Methoxy-2-naphthaldehyde 98%
6-Methoxy-2-naphthaldehyde
Vzorec: C₁₂H₁₀O₂
MW: 186,21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00081152
CAS číslo: 3453-33-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR