Tisk…

N-(2-Chloroethyl)benzamide 96%

Dodavatel: Apollo Scientific

OR900111-25G OR900111-5G
APOSOR900111-25GEA 5590 CZK
APOSOR900111-25G APOSOR900111-5G
N-(2-Chloroethyl)benzamide 96%
N-(2-Chloroethyl)benzamide
Vzorec: C₉H₁₀ClNO
Storage Temperature: Ambient
MDL Number: MFCD00000964
CAS číslo: 26385-07-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR