Tisk…

Methyl indoline-5-carboxylate 98%

Dodavatel: Apollo Scientific

OR900065-10G OR900065-1G OR900065-5G
APOSOR900065-10GEA 7440 CZK
APOSOR900065-10G APOSOR900065-1G APOSOR900065-5G
Methyl indoline-5-carboxylate 98%
Methyl indoline-5-carboxylate
Vzorec: C₁₀H₁₁NO₂
MW: 177,1998 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07371630
CAS číslo: 141452-01-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR