Tisk…

Aniline sulphate

Dodavatel: Apollo Scientific
Nebezpečí

OR8983-25G OR8983-5G OR8983-100G
APOSOR8983-25GEA 1240 CZK
APOSOR8983-25G APOSOR8983-5G APOSOR8983-100G
Aniline sulphate
Aniline sulphate
Vzorec: C₁₂H₁₆N₂O₄S
MW: 284.34 g/mol
Hustota: 1.377 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00013110
CAS číslo: 542-16-5
UN: 2811
ADR: 6.1 III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR