4-Nitro-1H-pyrazole-3-carboxylic acid 98%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
4-Nitro-3-pyrazolecarboxylic acid, 4-Nitropyrazole-3-carboxylic acid
OR8951-100G
APOSOR8951-100GEA
4570
CZK
APOSOR8951-100G
4-Nitro-1H-pyrazole-3-carboxylic acid 98%
4-Nitro-1H-pyrazole-3-carboxylic acid
Vzorec:
C₄H₃N₃O₄ MW: 157,085 g/mol Bod tání: 223…225 °C Storage Temperature: Ambient |
MDL Number:
MFCD00090899 CAS číslo: 5334-40-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...