Di-tert-butyl disulphide
Dodavatel: Apollo Scientific
Synonyms:
1,2-Di-tert-butyldisulphane, tert-Butyl disulphide
OR8920-100ML
OR8920-500ML
APOSOR8920-100MLEA
3540
CZK
APOSOR8920-100ML
APOSOR8920-500ML
Di-tert-butyl disulphide
Di-tert-butyl disulphide
Vzorec:
C₈H₁₈S₂ MW: 178,36 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00008838 CAS číslo: 110-06-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...