Methoxyhydroquinone
Dodavatel: Apollo Scientific
|
Varování
|
OR8900-1G
OR8900-5G
OR8900-10G
APOSOR8900-1GEA
622
CZK
APOSOR8900-1G
APOSOR8900-5G
APOSOR8900-10G
Methoxyhydroquinone
Methoxyhydroquinone
Vzorec:
C₇H₈O₃ Bod tání: 87…91 °C Storage Temperature: Ambient |
MDL Number:
MFCD00013971 CAS číslo: 824-46-4 EINECS: 212-530-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...