4-Amino-3-methylpyridine
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
4-Amino-3-picoline, 3-Methyl-4-pyridylamine, 3-Methylpyridin-4-amine
OR8895-1G
OR8895-25G
OR8895-5G
OR8895-100G
APOSOR8895-1GEA
0
CZK
APOSOR8895-1G
APOSOR8895-25G
APOSOR8895-5G
APOSOR8895-100G
4-Amino-3-methylpyridine
4-Amino-3-methylpyridine
Vzorec:
C₆H₈N₂ MW: 108,14 g/mol Bod tání: 106…107 °C Storage Temperature: Ambient |
MDL Number:
MFCD01704431 CAS číslo: 1990-90-5 EINECS: 217-872-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...