Tisk…

3-Vinylpyridine

Dodavatel: Apollo Scientific
Nebezpečí

OR8894-1G OR8894-5G OR8894-10G
APOSOR8894-1GEA 3740 CZK
APOSOR8894-1G APOSOR8894-5G APOSOR8894-10G
3-Vinylpyridine
3-Vinylpyridine
Vzorec: C₇H₇N
Bod varu: 85 °C/10mmHg
Hustota: 0,98 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD02178019
CAS číslo: 1121-55-7
UN: 3073
ADR: 6.1+3+8,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR