Isopropylboronic acid 98%
Dodavatel: Apollo Scientific
|
Varování
|
OR8880-100G
APOSOR8880-100GEA
4570
CZK
APOSOR8880-100G
Isopropylboronic acid 98%
Isopropylboronic acid
Vzorec:
C₃H₉BO₂ MW: 87.91 Bod tání: 95…100 °C Storage Temperature: Freezer |
MDL Number:
MFCD01319021 CAS číslo: 80041-89-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...