2-Amino-3-methoxypyridine 97%
Dodavatel: Apollo Scientific
|
Varování
|
OR8879-1G
OR8879-5G
APOSOR8879-1GEA
2570
CZK
APOSOR8879-1G
APOSOR8879-5G
2-Amino-3-methoxypyridine 97%
2-Amino-3-methoxypyridine
Vzorec:
C₆H₈N₂O Storage Temperature: Ambient |
MDL Number:
MFCD07374874 CAS číslo: 10201-71-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...