3-Bromopicolinic acid
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
3-Bromopyridine-2-carboxylic acid, 3-Bromo-2-pyridinecarboxylic acid
OR8877-1G
OR8877-5G
OR8877-25G
OR8877-100G
APOSOR8877-1GEA
622
CZK
APOSOR8877-1G
APOSOR8877-5G
APOSOR8877-25G
APOSOR8877-100G
3-Bromopicolinic acid
3-Bromopicolinic acid
Vzorec:
C₆H₄BrNO₂ MW: 202.01 Bod tání: 141…144 °C Hustota: 1.813 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD01320380 CAS číslo: 30683-23-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...