4-Nitroanthranilic acid 98+%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
2-Amino-4-nitrobenzoic acid
OR8876-100G
OR8876-25G
OR8876-500G
APOSOR8876-100GEA
1580
CZK
APOSOR8876-100G
APOSOR8876-25G
APOSOR8876-500G
4-Nitroanthranilic acid 98+%
4-Nitroanthranilic acid
Vzorec:
C₇H₆N₂O₄ Storage Temperature: Ambient |
MDL Number:
MFCD00007262 CAS číslo: 619-17-0 EINECS: 210-583-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...