Tisk…

2,3,5-Trichlorophenylboronic acid 98%

Dodavatel: Apollo Scientific

Synonyms: 2,3,5-Trichlorobenzeneboronic acid, Dihydroxy(2,3,5-trichlorophenyl)borane, (2,3,5-Trichlorophenyl)metaboric acid

OR8867-5G
APOSOR8867-5GEA 2150 CZK
APOSOR8867-5G
2,3,5-Trichlorophenylboronic acid 98%
2,3,5-Trichlorophenylboronic acid
Vzorec: C₆H₄BCl₃O₂
MW: 225,26 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01630856
CAS číslo: 212779-19-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR