5-Bromopyrimidin-2-ol
Dodavatel: Apollo Scientific
|
Varování
|
OR8863-100G
OR8863-25G
OR8863-5G
APOSOR8863-100GEA
9720
CZK
APOSOR8863-100G
APOSOR8863-25G
APOSOR8863-5G
5-Bromopyrimidin-2-ol
5-Bromopyrimidin-2-ol
Vzorec:
C₄H₃BrN₂O Storage Temperature: Ambient |
MDL Number:
MFCD00233952 CAS číslo: 214290-49-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...