(±)-Methyl-2-bromobutyrate
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
Methyl 2-bromobutyrate
OR8824-1KG
OR8824-500G
APOSOR8824-1KGEA
7380
CZK
APOSOR8824-1KG
APOSOR8824-500G
(±)-Methyl-2-bromobutyrate
(±)-Methyl-2-bromobutyrate
Vzorec:
C₅H₉BrO₂ Bod varu: 137…138 °C Hustota: 1,573 g/cm³ (25 °C) Bod vzplanutí: 68 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009666 CAS číslo: 3196-15-4 EINECS: 221-699-0 UN: 3265 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...