1-(6-Methylpyridin-2-yl)piperazine 98%
Dodavatel: Apollo Scientific
|
Varování
|
OR8754-1G
OR8754-5G
OR8754-250MG
APOSOR8754-1GEA
1870
CZK
APOSOR8754-1G
APOSOR8754-5G
APOSOR8754-250MG
1-(6-Methylpyridin-2-yl)piperazine 98%
1-(6-Methylpyridin-2-yl)piperazine
Vzorec:
C₁₀H₁₅N₃ MW: 177,25 g/mol Bod varu: 112…114 °C/0,1mmHg Storage Temperature: Ambient |
MDL Number:
MFCD01320891 CAS číslo: 55745-89-6 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...