Tisk…

D-Phenylalaninol

Dodavatel: Apollo Scientific
Nebezpečí

OR8744-100G OR8744-25G
APOSOR8744-100GEA 2910 CZK
APOSOR8744-100G APOSOR8744-25G
D-Phenylalaninol
D-Phenylalaninol
Vzorec: C₉H₁₃NO
Bod tání: 89…95 °C
Storage Temperature: Ambient
MDL Number: MFCD00064298
CAS číslo: 5267-64-1
UN: 3259
ADR: 8,III

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR