Tisk…

(S)-(+)-β-Aminophenethyl alcohol

Dodavatel: Apollo Scientific
Varování

OR8743-100G OR8743-25G OR8743-500G
APOSOR8743-100GEA 1290 CZK
APOSOR8743-100G APOSOR8743-25G APOSOR8743-500G
(S)-(+)-β-Aminophenethyl alcohol
(S)-(+)-β-Aminophenethyl alcohol
Vzorec: C₈H₁₁NO
Bod tání: 75…78 °C
Storage Temperature: Ambient
MDL Number: MFCD00064404
CAS číslo: 20989-17-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR