Tisk…

(R)-(-)-β-Aminophenethyl alcohol 95%

Dodavatel: Apollo Scientific
Varování

Synonyms: D-Phenylglycinol, (R)-(-)-2-Phenylglycinol

OR8742-100G OR8742-25G OR8742-5G
APOSOR8742-100GEA 4300 CZK
APOSOR8742-100G APOSOR8742-25G APOSOR8742-5G
(R)-(-)-β-Aminophenethyl alcohol 95%
(R)-(-)-β-Aminophenethyl alcohol
Vzorec: C₈H₁₁NO
MW: 137,18 g/mol
Bod tání: 75…78 °C
Storage Temperature: Ambient
MDL Number: MFCD00008062
CAS číslo: 56613-80-0
EINECS: 260-287-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR