(S)-(-)-4-Isopropyl-2-oxazolidinone
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
(4S)-(-)-Isopropyl-2-oxazolidinone, (4S)-(-)-4-Isopropyl-2-oxazolidinone, (S)-4-Isopropyl-2-oxazolidinone, (4S)-2-Oxo-4-(prop-2-yl)-1,3-oxazolidine, (4S)-4-(Prop-2-yl)-1,3-oxazolidin-2-one
OR8728-25G
OR8728-5G
APOSOR8728-25GEA
1490
CZK
APOSOR8728-25G
APOSOR8728-5G
(S)-(-)-4-Isopropyl-2-oxazolidinone
(S)-(-)-4-Isopropyl-2-oxazolidinone
Vzorec:
C₆H₁₁NO₂ MW: 129,16 g/mol Bod tání: 70…73 °C Storage Temperature: Ambient |
MDL Number:
MFCD00010847 CAS číslo: 17016-83-0 EINECS: 469-920-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...