(-)-4,5-Bis[hydroxy(diphenyl)methyl]-2,2-dimethyl-1,3-dioxolane 96%
Dodavatel: Apollo Scientific
Synonyms:
(4R,5R)-2,2-Dimethyl-alpha,alpha,alpha',alpha'-tetraphenyldioxolane-4,5-dimethanol, (R,R)-TADDOL, (4R,5R)-(-)-2,2-Dimethyl-α,α,α',α'-tetraphenyl-1,3-dioxolane-4,5-dimethanol, (-)-2,3-O-Isopropylidene-1,1,4,4-tetraphenyl-L-threitol
OR8725-25G
OR8725-5G
APOSOR8725-25GEA
6350
CZK
APOSOR8725-25G
APOSOR8725-5G
(-)-4,5-Bis[hydroxy(diphenyl)methyl]-2,2-dimethyl-1,3-dioxolane 96%
(-)-4,5-Bis[hydroxy(diphenyl)methyl]-2,2-dimethyl-1,3-dioxolane
Vzorec:
C₃₁H₃₀O₄ MW: 466,58 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00064467 CAS číslo: 93379-48-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...