Tisk…

4-(1-Pyrrolidinylmethyl)benzeneboronic acid pinacol ester 97%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 1-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl)pyrrolidine

OR8700-1G OR8700-250MG
APOSOR8700-1GEA 3110 CZK
APOSOR8700-1G APOSOR8700-250MG
4-(1-Pyrrolidinylmethyl)benzeneboronic acid pinacol ester 97%
4-(1-Pyrrolidinylmethyl)benzeneboronic acid pinacol ester
Vzorec: C₁₇H₂₆BNO₂
MW: 287,20 g/mol
Bod tání: 45…50 °C
Storage Temperature: Ambient
MDL Number: MFCD08690299
CAS číslo: 884507-39-5
UN: 2923
ADR: 8+6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR