Tisk…

1-Methylindole-4-boronic acid pinacol ester 97%

Dodavatel: Apollo Scientific
Varování

Synonyms: 1-Methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole

OR8699-1G OR8699-50MG OR8699-5G
APOSOR8699-1GEA 4070 CZK
APOSOR8699-1G APOSOR8699-50MG APOSOR8699-5G
1-Methylindole-4-boronic acid pinacol ester 97%
1-Methylindole-4-boronic acid pinacol ester
Vzorec: C₁₅H₂₀BNO₂
MW: 257.1400 g/mol
Bod tání: 95…96 °C
Storage Temperature: Freezer
MDL Number: MFCD08690266
CAS číslo: 898289-06-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR