2-(Phenoxymethyl)benzaldehyde 97%
Dodavatel: Apollo Scientific
|
Varování
|
OR8678-1G
OR8678-250MG
OR8678-50MG
APOSOR8678-1GEA
11610
CZK
APOSOR8678-1G
APOSOR8678-250MG
APOSOR8678-50MG
2-(Phenoxymethyl)benzaldehyde 97%
2-(Phenoxymethyl)benzaldehyde
Vzorec:
C₁₄H₁₂O₂ Bod varu: 180 °C/0,2mmHg Storage Temperature: Ambient |
MDL Number:
MFCD08690310 CAS číslo: 168551-49-3 UN: 2810 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...