Methyl-2-(methylsulphanyl)pyrimidine-5-carboxylate, Technical Grade
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
Methyl 2-methylthiopyrimidine-5-carboxylate, Methyl 2-(methylthio)pyrimidine-5-carboxylate
OR8662-1G
OR8662-250MG
OR8662-5G
APOSOR8662-1GEA
2070
CZK
APOSOR8662-1G
APOSOR8662-250MG
APOSOR8662-5G
Methyl-2-(methylsulphanyl)pyrimidine-5-carboxylate, Technical Grade
Methyl-2-(methylsulphanyl)pyrimidine-5-carboxylate
Vzorec:
C₇H₈N₂O₂S Bod tání: 96…97 °C Storage Temperature: Ambient |
MDL Number:
MFCD00194932 CAS číslo: 38275-41-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...