1-Methyl-1H-indole-4-carbonitrile 97%
Dodavatel: Apollo Scientific
|
Varování
|
OR8653-100MG
OR8653-5G
APOSOR8653-100MGEA
622
CZK
APOSOR8653-100MG
APOSOR8653-5G
1-Methyl-1H-indole-4-carbonitrile 97%
1-Methyl-1H-indole-4-carbonitrile
Vzorec:
C₁₀H₈N₂ Storage Temperature: Ambient |
MDL Number:
MFCD08690265 CAS číslo: 628711-58-0 UN: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...