3-Amino-1-methyl-1H-pyrazole-4-carboxaldehyde 97%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
3-Amino-1-methyl-1H-pyrazole-4-carbaldehyde
OR8638-250MG
APOSOR8638-250MGEA
7840
CZK
APOSOR8638-250MG
3-Amino-1-methyl-1H-pyrazole-4-carboxaldehyde 97%
3-Amino-1-methyl-1H-pyrazole-4-carboxaldehyde
Vzorec:
C₅H₇N₃O MW: 125,13 g/mol Bod tání: 133…134 °C Storage Temperature: Ambient |
MDL Number:
MFCD00460466 CAS číslo: 886851-66-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...