2-Aminonicotinonitrile
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
2-Aminopyridine-3-carbonitrile, 2-Amino-3-cyanopyridine
OR8628-1G
OR8628-5G
OR8628-10G
APOSOR8628-1GEA
622
CZK
APOSOR8628-1G
APOSOR8628-5G
APOSOR8628-10G
2-Aminonicotinonitrile
2-Aminonicotinonitrile
Vzorec:
C₆H₅N₃ MW: 119.13 Bod varu: 268 Bod tání: 132…136 °C Storage Temperature: Ambient |
MDL Number:
MFCD00128867 CAS číslo: 24517-64-4 UN: 3439 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...