4-Bromomethyl-2-cyanobiphenyl 95%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
4'-(Bromomethyl)-[1,1'-biphenyl]-2-carbonitrile
OR8619-500G
APOSOR8619-500GEA
1910
CZK
APOSOR8619-500G
4-Bromomethyl-2-cyanobiphenyl 95%
4-Bromomethyl-2-cyanobiphenyl
Vzorec:
C₁₄H₁₀BrN MW: 272,14 g/mol Bod tání: 125…128 °C Storage Temperature: Ambient |
MDL Number:
MFCD00671503 CAS číslo: 114772-54-2 EINECS: 425-280-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...