5-Bromo-2-chloro-4-methylpyridine
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
5-Bromo-2-chloro-4-picoline
OR8616-25G
OR8616-5G
OR8616-100G
APOSOR8616-25GEA
744
CZK
APOSOR8616-25G
APOSOR8616-5G
APOSOR8616-100G
5-Bromo-2-chloro-4-methylpyridine
5-Bromo-2-chloro-4-methylpyridine
Vzorec:
C₆H₅BrClN MW: 206.47 Bod tání: 25…30 °C Storage Temperature: Ambient |
MDL Number:
MFCD03427663 CAS číslo: 778611-64-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...