2-Bromo-5-chloro-3-nitropyridine 97%
Dodavatel: Apollo Scientific
OR8615-10G
OR8615-100G
OR8615-25G
APOSOR8615-10GEA
1990
CZK
APOSOR8615-10G
APOSOR8615-100G
APOSOR8615-25G
2-Bromo-5-chloro-3-nitropyridine 97%
2-Bromo-5-chloro-3-nitropyridine
Vzorec:
C₅H₂BrClN₂O₂ Storage Temperature: Ambient |
MDL Number:
MFCD07368982 CAS číslo: 75806-86-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...