Tisk…

5-Bromo-2-(1-pyrrolidinyl)pyridine

Dodavatel: Apollo Scientific
Varování

OR8602-1G OR8602-5G OR8602-25G
APOSOR8602-1GEA 622 CZK
APOSOR8602-1G APOSOR8602-5G APOSOR8602-25G
5-Bromo-2-(1-pyrrolidinyl)pyridine
5-Bromo-2-(1-pyrrolidinyl)pyridine
Vzorec: C₉H₁₁BrN₂
MW: 227,11 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07781213
CAS číslo: 210963-93-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR