Tisk…

5-Bromo-2-hydroxypyrimidine

Dodavatel: Apollo Scientific
Varování

Synonyms: 5-Bromopyrimidin-2(1H)-one, 5-Bromo-2-pyrimidinone

OR8599-1G OR8599-25G OR8599-5G
APOSOR8599-1GEA 622 CZK
APOSOR8599-1G APOSOR8599-25G APOSOR8599-5G
5-Bromo-2-hydroxypyrimidine
5-Bromo-2-hydroxypyrimidine
Vzorec: C₄H₃BrN₂O
MW: 174,98 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01318956
CAS číslo: 38353-06-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR