Tisk…

(±)-Nipecotic acid 98%

Dodavatel: Apollo Scientific
Varování

Synonyms: H-DL-Nip-OH, (±)-3-Piperidinecarboxylic acid, Piperidine-3-carboxylic acid, Nipecotic acid

OR8589-100G OR8589-1KG OR8589-500G
APOSOR8589-100GEA 3540 CZK
APOSOR8589-100G APOSOR8589-1KG APOSOR8589-500G
(±)-Nipecotic acid 98%
(±)-Nipecotic acid
Vzorec: C₆H₁₁NO₂
MW: 129,16 g/mol
Bod tání: 261 °C
Storage Temperature: Ambient
MDL Number: MFCD00005992
CAS číslo: 498-95-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR